3-Fluor-4-(isopropylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

PC3587-1G
APOSPC3587-1GEA 248 EUR
APOSPC3587-1G
3-Fluor-4-(isopropylcarbamoyl)phenylboronsäure 98%
3-Fluor-4-(isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Schmelzpunkt: 137…139 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08436040
CAS-Nummer: 874289-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR