(1R)-1-(Trifluoromethyl)ethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R)-2-Amino-1,1,1-trifluoropropane hydrochloride, (R)-1,1,1-Trifluoropropan-2-amine hydrochloride, (2R)-1,1,1-Trifluoropropan-2-amine hydrochloride

PC3589-1G PC3589-250MG
APOSPC3589-1GEA 59.7 EUR
APOSPC3589-1G APOSPC3589-250MG
(1R)-1-(Trifluoromethyl)ethylamine hydrochloride
(1R)-1-(Trifluoromethyl)ethylamine hydrochloride
Formel: C₃H₆F₃N·ClH
Molecular Weight: 149.543 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779095
CAS-Nummer: 177469-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR