2-Brom-3-chlorbenzotrifluorid 98%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-1-chloro-3-(trifluoromethyl)benzene

PC3593-5G
APOSPC3593-5GEA 89.9 EUR
APOSPC3593-5G
2-Brom-3-chlorbenzotrifluorid 98%
2-Brom-3-chlorbenzotrifluorid
Formel: C₇H₃BrClF₃
Molecular Weight: 259,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673988
CAS-Nummer: 384-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR