3-Methyl-5-(trifluoromethoxy)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC3602-5G PC3602-1G PC3602-250MG
APOSPC3602-5GEA 171 EUR
APOSPC3602-5G APOSPC3602-1G APOSPC3602-250MG
3-Methyl-5-(trifluoromethoxy)benzaldehyde
3-Methyl-5-(trifluoromethoxy)benzaldehyde
Formel: C₉H₇F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08741401
CAS-Nummer: 1000339-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR