3-Methyl-5-(trifluormethoxy)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3604-1G
PC3604-5G
APOSPC3604-1GEA
69.6
EUR
APOSPC3604-1G
APOSPC3604-5G
3-Methyl-5-(trifluormethoxy)benzylbromid 98%
3-Methyl-5-(trifluormethoxy)benzylbromid
Formel:
C₉H₈BrF₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741403 CAS-Nummer: 916420-55-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...