3-Methyl-5-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC3604-1G PC3604-5G
APOSPC3604-1GEA 69.6 EUR
APOSPC3604-1G APOSPC3604-5G
3-Methyl-5-(trifluormethoxy)benzylbromid 98%
3-Methyl-5-(trifluormethoxy)benzylbromid
Formel: C₉H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD08741403
CAS-Nummer: 916420-55-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR