3-Methyl-5-(trifluormethoxy)phenylessigsäure 97%

Lieferant: Apollo Scientific
Achtung

PC3606-1G
APOSPC3606-1GEA 260 EUR
APOSPC3606-1G
3-Methyl-5-(trifluormethoxy)phenylessigsäure 97%
3-Methyl-5-(trifluormethoxy)phenylessigsäure
Formel: C₉H₉F₃O₃
Molecular Weight: 234,17 g/mol
Schmelzpunkt: 52…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08741405
CAS-Nummer: 1000339-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR