3-Fluor-2-methoxy-5-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Fluoro-6-methoxypyridin-3-yl)boronic acid, 3-Fluoro-2-methoxypyridine-5-boronic acid

PC3612-1G
APOSPC3612-1GEA 75.4 EUR
APOSPC3612-1G
3-Fluor-2-methoxy-5-pyridinboronsäure
3-Fluor-2-methoxy-5-pyridinboronsäure
Formel: C₆H₇BFNO₃
Molecular Weight: 170.9341 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04112520
CAS-Nummer: 856250-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR