2-Chlor-3-fluor-5-pyridinboronsäure 95+%

Lieferant: Apollo Scientific

Synonyme: (6-Chloro-5-fluoropyridin-3-yl)boronic acid

PC3616-1G
APOSPC3616-1GEA 77 EUR
APOSPC3616-1G
2-Chlor-3-fluor-5-pyridinboronsäure 95+%
2-Chlor-3-fluor-5-pyridinboronsäure
Formel: C₅H₄BClFNO₂
Molecular Weight: 175,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112500
CAS-Nummer: 1072946-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR