2-Chlor-5-fluor-4-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-fluoropyridine-4-boronic Acid

PC3618-5G
APOSPC3618-5GEA 190 EUR
APOSPC3618-5G
2-Chlor-5-fluor-4-pyridinboronsäure
2-Chloro-5-fluoro-4-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175.35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04972404
CAS-Nummer: 951677-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR