2-Fluor-6-methyl-5-pyridinboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-6-methylpyridine-5-boronic acid, (6-Fluoro-2-methylpyridin-3-yl)boronic acid

PC3619-5G
APOSPC3619-5GEA 106 EUR
APOSPC3619-5G
2-Fluor-6-methyl-5-pyridinboronsäure 97%
2-Fluor-6-methyl-5-pyridinboronsäure
Formel: C₆H₇BFNO₂
Molecular Weight: 154,9400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411561
CAS-Nummer: 904326-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR