4-(Butylcarbamoyl)-3-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific

PC3622-1G
APOSPC3622-1GEA 198 EUR
APOSPC3622-1G
4-(Butylcarbamoyl)-3-fluorphenylboronsäure 98%
4-(Butylcarbamoyl)-3-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Schmelzpunkt: 130…133 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689527
CAS-Nummer: 874289-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR