5-Fluor-3-pyridinboronsäure 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid

PC3624-1G PC3624-5G PC3624-25G
APOSPC3624-1GEA 29 EUR
APOSPC3624-1G APOSPC3624-5G APOSPC3624-25G
5-Fluor-3-pyridinboronsäure 95+%
5-Fluoro-3-pyridineboronic acid
Formel: C₅H₅BFNO₂
Molecular Weight: 140.91 g/mol
Schmelzpunkt: 233…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368243
CAS-Nummer: 872041-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR