4-Benzylcarbamoyl-3-fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
Synonyme:
(4-(Benzylcarbamoyl)-3-fluorophenyl)boronic acid, 4-(Benzylcarbamoyl)-3-fluorophenylboronic acid
PC3625-1G
APOSPC3625-1GEA
241
EUR
APOSPC3625-1G
4-Benzylcarbamoyl-3-fluorphenylboronsäure 97%
4-Benzylcarbamoyl-3-fluorphenylboronsäure
Formel:
C₁₄H₁₃BFNO₃ Molecular Weight: 273,0700 g/mol Schmelzpunkt: 113…116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08689501 CAS-Nummer: 874289-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...