6-Fluoro-4-hydroxyquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC3627-500MG
APOSPC3627-500MGEA
21.9
EUR
APOSPC3627-500MG
6-Fluoro-4-hydroxyquinoline-3-carboxylic acid
6-Fluoro-4-hydroxyquinoline-3-carboxylic acid
Formel:
C₁₀H₆FNO₃ Schmelzpunkt: 298…300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00973836 CAS-Nummer: 343-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...