2-Brom-6-fluor-4-nitroanisol 97%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-fluoro-4-methoxynitrobenzene

PC3628-5G
APOSPC3628-5GEA 326 EUR
APOSPC3628-5G
2-Brom-6-fluor-4-nitroanisol 97%
2-Brom-6-fluor-4-nitroanisol
Formel: C₇H₅BrFNO₃
Molecular Weight: 250,02 g/mol
Schmelzpunkt: 70…71 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08062383
CAS-Nummer: 875664-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR