5-(Trifluormethoxy)isatin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3629-5G
APOSPC3629-5GEA
66.7
EUR
APOSPC3629-5G
5-(Trifluormethoxy)isatin 97%
5-(Trifluormethoxy)isatin
Formel:
C₉H₄F₃NO₃ Schmelzpunkt: 170…172 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00192524 CAS-Nummer: 169037-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...