4-Fluor-3-hydroxybenzonitril 97%

Lieferant: Apollo Scientific

Synonyme: 4-Fluor-3-hydroxybenzoesäurenitril

PC3631-1G
APOSPC3631-1GEA 21.9 EUR
APOSPC3631-1G
4-Fluor-3-hydroxybenzonitril 97%
4-Fluor-3-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137,11 g/mol
MDL: MFCD08689519
CAS-Nummer: 186590-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR