4-(4-Fluorbenzoyl)piperidin Hydrochlorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluorophenyl)(piperidin-4-yl)methanone hydrochloride
PC3634-25G
APOSPC3634-25GEA
0
EUR
APOSPC3634-25G
4-(4-Fluorbenzoyl)piperidin Hydrochlorid 97%
4-(4-Fluorbenzoyl)piperidin Hydrochlorid
Formel:
C₁₂H₁₅ClFNO Molecular Weight: 243.71 g/mol Schmelzpunkt: 222…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044912 CAS-Nummer: 25519-78-2 EINECS: 247-070-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...