3-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid

Lieferant: Apollo Scientific

PC3651-1G
APOSPC3651-1GEA 176 EUR
APOSPC3651-1G
3-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid
3-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid
Formel: C₁₂H₉BFNO₅
Molecular Weight: 277,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878354
CAS-Nummer: 1072945-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR