3-Fluoro-4-(methylsulphonyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

PC3656-250MG
APOSPC3656-250MGEA 0 EUR
APOSPC3656-250MG
3-Fluoro-4-(methylsulphonyl)benzeneboronic acid
3-Fluoro-4-(methylsulphonyl)benzeneboronic acid
Formel: C₇H₈BFO₄S
Schmelzpunkt: 103…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09878323
CAS-Nummer: 648904-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR