1-(2-Fluorophenyl)-1H-pyrazole-4-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

PC3657-1G
APOSPC3657-1GEA 176 EUR
APOSPC3657-1G
1-(2-Fluorophenyl)-1H-pyrazole-4-carboxaldehyde
1-(2-Fluorophenyl)-1H-pyrazole-4-carboxaldehyde
Formel: C₁₀H₇FN₂O
Molecular Weight: 190,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864518
CAS-Nummer: 1015845-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR