Ethyl 2-methyl-6-(trifluoromethyl)nicotinate 95+%

Lieferant: Apollo Scientific

PC3721-10G PC3721-5G PC3721-1G PC3721-250MG
APOSPC3721-10GEA 0 EUR
APOSPC3721-10G APOSPC3721-5G APOSPC3721-1G APOSPC3721-250MG
Ethyl 2-methyl-6-(trifluoromethyl)nicotinate 95+%
Ethyl-2-methyl-6-(trifluoromethyl)nicotinate
Formel: C₁₀H₁₀F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10688283
CAS-Nummer: 380355-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR