3-Fluorbenzylisothiocyanat 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3721M-1G
PC3721M-5G
PC3721M-250MG
APOSPC3721M-1GEA
63.8
EUR
APOSPC3721M-1G
APOSPC3721M-5G
APOSPC3721M-250MG
3-Fluorbenzylisothiocyanat 97%
3-Fluorbenzylisothiocyanat
Formel:
C₈H₆FNS Molecular Weight: 167.21 g/mol Siedepunkt: 138 °C/16mmHg Dichte: 1.22 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00060406 CAS-Nummer: 63351-94-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...