2-[(4-Chlorbenzyl)oxy]-6-fluorbenzonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3728E-1G
APOSPC3728E-1GEA
60.9
EUR
APOSPC3728E-1G
2-[(4-Chlorbenzyl)oxy]-6-fluorbenzonitril 97%
2-[(4-Chlorbenzyl)oxy]-6-fluorbenzonitril
Formel:
C₁₄H₉ClFNO Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068205 CAS-Nummer: 175204-10-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...