6-Fluoro-4-chromanone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluorochroman-4-one

PC3728P-1G
APOSPC3728P-1GEA 21.9 EUR
APOSPC3728P-1G
6-Fluoro-4-chromanone 98%
6-Fluoro-4-chromanone
Formel: C₉H₇FO₂
Molecular Weight: 166,15 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038654
CAS-Nummer: 66892-34-0
EINECS: 266-511-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR