4-Fluorzimtsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(p-Fluorphenyl)propensäure, 4-Fluorcinnamsäure, 3-(4-Fluorphenyl)propensäure
PC3732-100G
PC3732-250G
PC3732-1KG
PC3732-500G
APOSPC3732-100GEA
33.5
EUR
APOSPC3732-100G
APOSPC3732-250G
APOSPC3732-1KG
APOSPC3732-500G
4-Fluorzimtsäure 99%
4-Fluorzimtsäure
Formel:
C₉H₇FO₂ Schmelzpunkt: 208…210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004395 CAS-Nummer: 459-32-5 EINECS: 207-288-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...