6-Fluoro-6-deoxy-D-galactopyranose 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Deoxy-6-fluoro-D-galactopyranose

PC3738J-25MG
APOSPC3738J-25MGEA 125 EUR
APOSPC3738J-25MG
6-Fluoro-6-deoxy-D-galactopyranose 97%
6-Fluoro-6-deoxy-D-galactopyranose
Formel: C₆H₁₁FO₅
Molecular Weight: 182,15 g/mol
Schmelzpunkt: 161…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063304
CAS-Nummer: 18961-68-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR