4-Brom-2-chlor-6-fluoranisol

Lieferant: Apollo Scientific
Achtung

PC3746-1G
APOSPC3746-1GEA 46.4 EUR
APOSPC3746-1G
4-Brom-2-chlor-6-fluoranisol
4-Brom-2-chlor-6-fluoranisol
Formel: C₇H₅BrClFO
Molecular Weight: 239,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631477
CAS-Nummer: 261762-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR