6-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-2-(trifluoromethyl)quinolin-4-ol

PC3756X-1G
APOSPC3756X-1GEA 30.6 EUR
APOSPC3756X-1G
6-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
6-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline
Formel: C₁₀H₅F₄NO
Molecular Weight: 231,15 g/mol
Schmelzpunkt: 254…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276582
CAS-Nummer: 31009-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR