8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3756Y-10G
APOSPC3756Y-10GEA
212
EUR
APOSPC3756Y-10G
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline
Formel:
C₁₀H₅F₄NO Schmelzpunkt: 110…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00276580 CAS-Nummer: 31009-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...