1H,1H-Heptafluorobutylnonafluorobutanesulphonate 97%

Lieferant: Apollo Scientific
Gefahr

PC3762-5G
APOSPC3762-5GEA 575 EUR
APOSPC3762-5G
1H,1H-Heptafluorobutylnonafluorobutanesulphonate 97%
1H,1H-Heptafluorobutylnonafluorobutanesulphonate
Formel: C₈H₂F₁₆O₃S
Molecular Weight: 482,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04038309
CAS-Nummer: 883499-32-9
UN: 2927
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR