4-(4-Fluor-2-nitrophenyl)morpholin

Lieferant: Apollo Scientific
Achtung

PC3765-500MG
APOSPC3765-500MGEA 21.9 EUR
APOSPC3765-500MG
4-(4-Fluor-2-nitrophenyl)morpholin
4-(4-Fluor-2-nitrophenyl)morpholin
Formel: C₁₀H₁₁FN₂O₃
Molecular Weight: 226,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798369
CAS-Nummer: 238418-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR