2-Amino-6-(trifluoromethylthio)benzothiazole
Lieferant: Apollo Scientific
|
Achtung
|
PC3767-100MG
APOSPC3767-100MGEA
0
EUR
APOSPC3767-100MG
2-Amino-6-(trifluoromethylthio)benzothiazole
2-Amino-6-(trifluoromethylthio)benzothiazole
Formel:
C₈H₅F₃N₂S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665748 CAS-Nummer: 326-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...