1,2-Dibrom-1,1-dichlor-2,2-difluorethan 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Dibromo-1,1-dichlorodifluoroethane
PC3783-25G
APOSPC3783-25GEA
499
EUR
APOSPC3783-25G
1,2-Dibrom-1,1-dichlor-2,2-difluorethan 97%
1,2-Dibrom-1,1-dichlor-2,2-difluorethan
Formel:
C₂Br₂Cl₂F₂ Molecular Weight: 292,73 g/mol Siedepunkt: 117 °C Schmelzpunkt: 46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039319 CAS-Nummer: 558-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...