2-Bromo-4-fluoro-5-methylaniline 97%

Lieferant: Apollo Scientific
Achtung

PC3785-5G
APOSPC3785-5GEA 87 EUR
APOSPC3785-5G
2-Bromo-4-fluoro-5-methylaniline 97%
2-Bromo-4-fluoro-5-methylaniline
Formel: C₇H₇BrFN
Molecular Weight: 204.04 g/mol
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11042843
CAS-Nummer: 1065076-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR