Poly(vinylidene fluoride)

Lieferant: Apollo Scientific
Achtung

PC37872-100G PC37872-25G PC37872-5G
APOSPC37872-100GEA 444 EUR
APOSPC37872-100G APOSPC37872-25G APOSPC37872-5G
Poly(vinylidene fluoride)
Poly(vinylidene fluoride)
Formel: (-CH₂CF₂-)n
Schmelzpunkt: 155…165 °C
Dichte: 1.74 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00084470
CAS-Nummer: 24937-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR