2,2-Difluoroethyl trifluoromethanesulphonate 98%

Lieferant: Apollo Scientific
Gefahr

PC3793-5G PC3793-1G PC3793-25G
APOSPC3793-5GEA 24.8 EUR
APOSPC3793-5G APOSPC3793-1G APOSPC3793-25G
2,2-Difluoroethyl trifluoromethanesulphonate 98%
2,2-Difluoroethyl trifluoromethanesulphonate
Formel: C₃H₃F₅O₃S
Siedepunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093342
CAS-Nummer: 74427-22-8
UN: 2927
ADR: 6.1+8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR