Dimethyl threo-2-bromo-3-fluorosuccinate
Lieferant: Apollo Scientific
PC3795-100MG
APOSPC3795-100MGEA
170
EUR
APOSPC3795-100MG
Dimethyl threo-2-bromo-3-fluorosuccinate
Dimethyl threo-2-bromo-3-fluorosuccinate
Formel:
C₆H₈BrFO₄ Molecular Weight: 243,028 g/mol Dichte: 1,3328 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093313 CAS-Nummer: 36875-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...