3-Fluor-4-methoxybenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3799-1G
PC3799-5G
APOSPC3799-1GEA
33.5
EUR
APOSPC3799-1G
APOSPC3799-5G
3-Fluor-4-methoxybenzylalkohol 98%
3-Fluor-4-methoxybenzylalkohol
Formel:
C₈H₉FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070642 CAS-Nummer: 96047-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...