4-Brom-5-fluor-2-nitroanilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3817-500MG
APOSPC3817-500MGEA
21.9
EUR
APOSPC3817-500MG
4-Brom-5-fluor-2-nitroanilin 97%
4-Brom-5-fluor-2-nitroanilin
Formel:
C₆H₄BrFN₂O₂ Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101433 CAS-Nummer: 153505-36-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...