4'-Fluor-2'-methoxyacetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluoro-2-methoxyphenyl)ethanone

PC3817C-1G
APOSPC3817C-1GEA 21.9 EUR
APOSPC3817C-1G
4'-Fluor-2'-methoxyacetophenon 98%
4'-Fluor-2'-methoxyacetophenon
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
Schmelzpunkt: 51…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061143
CAS-Nummer: 51788-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR