3-Fluoro-4-methylbenzamide oxime 97%

Lieferant: Apollo Scientific

PC3822G-1G PC3822G-5G
APOSPC3822G-1GEA 0 EUR
APOSPC3822G-1G APOSPC3822G-5G
3-Fluoro-4-methylbenzamide oxime 97%
3-Fluoro-4-methylbenzamide oxime
Formel: C₈H₉FN₂O
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08461808
CAS-Nummer: 238742-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR