2-Fluor-3-methylbenzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-2-fluoro-3-methylbenzene
PC3823D-1G
PC3823D-5G
PC3823D-25G
APOSPC3823D-1GEA
21.9
EUR
APOSPC3823D-1G
APOSPC3823D-5G
APOSPC3823D-25G
2-Fluor-3-methylbenzylbromid 98%
2-Fluor-3-methylbenzylbromid
Formel:
C₈H₈BrF Siedepunkt: 168 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042470 CAS-Nummer: 151412-12-3 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...