(+/-)-1-(4-Fluorophenyl)ethyl isothiocyanate 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Fluoro-alpha-methylbenzyl isothiocyanate
PC3823G-1G
APOSPC3823G-1GEA
111
EUR
APOSPC3823G-1G
(+/-)-1-(4-Fluorophenyl)ethyl isothiocyanate 97%
(+/-)-1-(4-Fluorophenyl)ethyl isothiocyanate
Formel:
C₉H₈FNS Molecular Weight: 181,23 g/mol Siedepunkt: 109…110 °C/0,6mmHg Dichte: 1,15 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041105 CAS-Nummer: 182565-27-1 UN: 2922 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...