Methyl-2-hydroxy-3,3,3-trifluoropropanoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3,3,3-trifluoro-DL-lactate
PC3844-1G
APOSPC3844-1GEA
89.9
EUR
APOSPC3844-1G
Methyl-2-hydroxy-3,3,3-trifluoropropanoate 95%
Methyl-2-hydroxy-3,3,3-trifluoropropanoate
Formel:
C₄H₅F₃O₃ Molecular Weight: 158,08 g/mol Siedepunkt: 60…64 °C/20mmHg Schmelzpunkt: 59…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093316 CAS-Nummer: 93496-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...