4'-Fluor-2'-nitroacetanilid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Fluoro-2-nitrophenyl)acetamide

PC3852-25G
APOSPC3852-25GEA 132 EUR
APOSPC3852-25G
4'-Fluor-2'-nitroacetanilid 98%
4'-Fluor-2'-nitroacetanilid
Formel: C₈H₇FN₂O₃
Molecular Weight: 179,22 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031517
CAS-Nummer: 448-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR