3-Cyano-5-fluorobenzoic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3859-5G
APOSPC3859-5GEA
95.7
EUR
APOSPC3859-5G
3-Cyano-5-fluorobenzoic acid 98%
3-Cyano-5-fluorobenzoic acid
Formel:
C₈H₄FNO₂ Schmelzpunkt: 171…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663717 CAS-Nummer: 327056-74-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...