Kaliumperfluor-1-butansulfonat
Lieferant: Apollo Scientific
|
Achtung
|
PC3874-500G
APOSPC3874-500GEA
142
EUR
APOSPC3874-500G
Kaliumperfluor-1-butansulfonat
Kaliumperfluor-1-butansulfonat
Additive for polymers. Soluble in water, acetone, methanol & ethyl acetate
Formel:
C₄F₉KO₃S Schmelzpunkt: 300 °C (Zersetzung) Dichte: 0,69 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007515 CAS-Nummer: 29420-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...