2',4',6'-Trichloracetanilid
Lieferant: Apollo Scientific
|
Achtung
|
PC3888-1G
APOSPC3888-1GEA
118
EUR
APOSPC3888-1G
2',4',6'-Trichloracetanilid
2',4',6'-Trichloracetanilid
Formel:
C₈H₆Cl₃NO Schmelzpunkt: 152…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093332 CAS-Nummer: 363-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...